Tisk…

(3-Methylisoxazol-5-yl)acetonitrile

Dodavatel: Apollo Scientific
Varování

OR451424-1G OR451424-5G
APOSOR451424-1GEA 4960 CZK
APOSOR451424-1G APOSOR451424-5G
(3-Methylisoxazol-5-yl)acetonitrile
(3-Methylisoxazol-5-yl)acetonitrile
Vzorec: C₆H₆N₂O
MW: 122,125 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17078885
CAS číslo: 288318-31-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR