4-Bromo-5-methylthiophene-2-carbaldehyde
Dodavatel: Apollo Scientific
|
Varování
|
OR451423-1G
OR451423-5G
APOSOR451423-1GEA
2150
CZK
APOSOR451423-1G
APOSOR451423-5G
4-Bromo-5-methylthiophene-2-carbaldehyde
4-Bromo-5-methylthiophene-2-carbaldehyde
Vzorec:
C₆H₅BrOS MW: 205.072 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD01118659 CAS číslo: 29421-75-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...