Tisk…

(2Z)-1-(4-Chlorophenyl)-3-(dimethylamino)prop-2-en-1-one

Dodavatel: Apollo Scientific
Varování

OR451413-1G OR451413-5G
APOSOR451413-1GEA 992 CZK
APOSOR451413-1G APOSOR451413-5G
(2Z)-1-(4-Chlorophenyl)-3-(dimethylamino)prop-2-en-1-one
(2Z)-1-(4-Chlorophenyl)-3-(dimethylamino)prop-2-en-1-one
Vzorec: C₁₁H₁₂ClNO
Storage Temperature: Ambient
MDL Number: MFCD00172116
CAS číslo: 28587-05-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR