Tisk…

Methyl 1-(chloromethyl)-1H-pyrazole-3-carboxylate

Dodavatel: Apollo Scientific
Varování

OR451412-1G OR451412-5G
APOSOR451412-1GEA 5390 CZK
APOSOR451412-1G APOSOR451412-5G
Methyl 1-(chloromethyl)-1H-pyrazole-3-carboxylate
Methyl 1-(chloromethyl)-1H-pyrazole-3-carboxylate
Vzorec: C₆H₇ClN₂O₂
MW: 174,585 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03423395
CAS číslo: 1003293-40-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR