4-Chloro-5-methyl-3-nitro-1H-pyrazole
Dodavatel: Apollo Scientific
|
Varování
|
OR451410-1G
OR451410-5G
APOSOR451410-1GEA
1660
CZK
APOSOR451410-1G
APOSOR451410-5G
4-Chloro-5-methyl-3-nitro-1H-pyrazole
4-Chloro-5-methyl-3-nitro-1H-pyrazole
Vzorec:
C₄H₄ClN₃O₂ MW: 161,546 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00195034 CAS číslo: 512810-26-3 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...