3-Iodophenacyl bromide
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-Bromo-1-(3-iodophenyl)ethanone, 2-Bromo-1-(3-iodophenyl)ethan-1-one
OR45138-1G
OR45138-250MG
APOSOR45138-1GEA
14490
CZK
APOSOR45138-1G
APOSOR45138-250MG
3-Iodophenacyl bromide
3-Iodophenacyl bromide
Vzorec:
C₈H₆BrIO MW: 324,941 g/mol Bod tání: 62…65 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06666937 CAS číslo: 61858-38-6 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...