2-Bromo-4-iodo-6-methylanisole
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-Bromo-5-iodo-2-methoxy-3-methylbenzene, 2-Bromo-4-iodo-6-methylphenyl methyl ether, 3-Bromo-5-iodo-2-methoxytoluene
OR45135-1G
OR45135-250MG
OR45135-5G
OR45135-25G
APOSOR45135-1GEA
6620
CZK
APOSOR45135-1G
APOSOR45135-250MG
APOSOR45135-5G
APOSOR45135-25G
2-Bromo-4-iodo-6-methylanisole
2-Bromo-4-iodo-6-methylanisole
Vzorec:
C₈H₈BrIO MW: 326,957 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD27937157 CAS číslo: 1820675-25-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...