α-Furfuryl octanoate 98+%
Dodavatel: Apollo Scientific
|
Varování
|
OR451301-100G
OR451301-25G
OR451301-500G
APOSOR451301-100GEA
17860
CZK
APOSOR451301-100G
APOSOR451301-25G
APOSOR451301-500G
α-Furfuryl octanoate 98+%
α-Furfuryl octanoate
Vzorec:
C₁₃H₂₀O₃ MW: 224,296 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00209510 CAS číslo: 39252-03-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...