Tisk…
Ethyl 3-oxo-3-(tetrahydro-2H-pyran-4-yl)propanoate
Dodavatel: Apollo Scientific
Synonyms: Ethyl 3-Oxo-3-(4-tetrahydropyranyl)propanoate
OR451223-1G OR451223-250MG OR451223-5G
APOSOR451223-1GEA 2360 CZK
APOSOR451223-1G APOSOR451223-250MG APOSOR451223-5G
Ethyl 3-oxo-3-(tetrahydro-2H-pyran-4-yl)propanoate
Ethyl 3-oxo-3-(tetrahydro-2H-pyran-4-yl)propanoate
Vzorec: C₁₀H₁₆O₄
MW: 200.232 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115573
CAS číslo: 856414-68-1