Tisk…

3-Bromo-2-iodoanisole

Dodavatel: Apollo Scientific
Varování

OR45121-1G OR45121-5G OR45121-25G
APOSOR45121-1GEA 953 CZK
APOSOR45121-1G APOSOR45121-5G APOSOR45121-25G
3-Bromo-2-iodoanisole
3-Bromo-2-iodoanisole
Vzorec: C₇H₆BrIO
Storage Temperature: Ambient
MDL Number: MFCD18089327
CAS číslo: 450412-22-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR