Tisk…

1-Cyclopentylpiperidin-4-amine dihydrochloride 98%

Dodavatel: Apollo Scientific

OR451198-5G OR451198-10G OR451198-1G
APOSOR451198-5GEA 20700 CZK
APOSOR451198-5G APOSOR451198-10G APOSOR451198-1G
1-Cyclopentylpiperidin-4-amine dihydrochloride 98%
1-Cyclopentylpiperidin-4-amine dihydrochloride
Vzorec: C₁₀H₂₀N₂·2ClH
MW: 241,201 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09064935
CAS číslo: 952201-42-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR