Tisk…

(2R)-2-Amino-2-cyclopropylethan-1-ol hydrochloride 95%

Dodavatel: Apollo Scientific
Varování

OR451119-1G OR451119-250MG
APOSOR451119-1GEA 15050 CZK
APOSOR451119-1G APOSOR451119-250MG
(2R)-2-Amino-2-cyclopropylethan-1-ol hydrochloride 95%
(2R)-2-Amino-2-cyclopropylethan-1-ol hydrochloride
Vzorec: C₅H₁₁NO·ClH
MW: 137,608 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28975219
CAS číslo: 1270290-36-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR