Tisk…

N-Cyclopropyl-4-chloro-benzylamine 95%

Dodavatel: Apollo Scientific

OR451115-250MG OR451115-1G OR451115-5G
APOSOR451115-250MGEA 5200 CZK
APOSOR451115-250MG APOSOR451115-1G APOSOR451115-5G
N-Cyclopropyl-4-chloro-benzylamine 95%
N-Cyclopropyl-4-chloro-benzylamine
Vzorec: C₁₀H₁₂ClN
Storage Temperature: Ambient
MDL Number: MFCD07408125
CAS číslo: 19271-24-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR