1-Benzyl-3-azetidinamine dihydrochloride 95%
Dodavatel: Apollo Scientific
OR451024-1G
OR451024-250MG
APOSOR451024-1GEA
20700
CZK
APOSOR451024-1G
APOSOR451024-250MG
1-Benzyl-3-azetidinamine dihydrochloride 95%
1-Benzyl-3-azetidinamine dihydrochloride
Vzorec:
C₁₀H₁₄N₂·2ClH MW: 235,153 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16293358 CAS číslo: 1307683-81-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...