Tisk…

3-(Chloromethyl)-5-[(morpholin-4-yl)methyl]isoxazole

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-{[3-(Chloromethyl)isoxazol-5-yl]methyl}morpholine

OR45060-1G OR45060-250MG
APOSOR45060-1GEA 7380 CZK
APOSOR45060-1G APOSOR45060-250MG
3-(Chloromethyl)-5-[(morpholin-4-yl)methyl]isoxazole
3-(Chloromethyl)-5-[(morpholin-4-yl)methyl]isoxazole
Vzorec: C₉H₁₃ClN₂O₂
MW: 216,665 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11841078
CAS číslo: 953408-97-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR