Tisk…

2-Phenylacetyl isocyanate

Dodavatel: Apollo Scientific

OR450129-1G OR450129-250MG
APOSOR450129-1GEA 46620 CZK
APOSOR450129-1G APOSOR450129-250MG
2-Phenylacetyl isocyanate
2-Phenylacetyl isocyanate

May form small amount of sediment

Vzorec: C₉H₇NO₂
Storage Temperature: Freezer
MDL Number: MFCD16301042
CAS číslo: 4461-27-2
UN: 2206
ADR: 6.1,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR