Tisk…

5-Chloro-[1,2,4]triazolo[1,5-a]pyrazine

Dodavatel: Apollo Scientific

OR450116-1G OR450116-250MG
APOSOR450116-1GEA 56870 CZK
APOSOR450116-1G APOSOR450116-250MG
5-Chloro-[1,2,4]triazolo[1,5-a]pyrazine
5-Chloro-[1,2,4]triazolo[1,5-a]pyrazine
Vzorec: C₅H₃ClN₄
Storage Temperature: Refrigerator
MDL Number: MFCD27996718
CAS číslo: 42399-82-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR