Tisk…

4-Amino-[1,2,4]triazolo[5,1-c][1,2,4]triazine-3-carbonitrile

Dodavatel: Apollo Scientific

OR450110-1G OR450110-250MG
APOSOR450110-1GEA 14490 CZK
APOSOR450110-1G APOSOR450110-250MG
4-Amino-[1,2,4]triazolo[5,1-c][1,2,4]triazine-3-carbonitrile
4-Amino-[1,2,4]triazolo[5,1-c][1,2,4]triazine-3-carbonitrile
Vzorec: C₅H₃N₇
Storage Temperature: Refrigerator
MDL Number: MFCD18804243
CAS číslo: 61033-27-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR