6-Methylbicyclo[4.1.0]heptan-2-one 95%
Dodavatel: Apollo Scientific
OR45011-1G
OR45011-250MG
OR45011-5G
APOSOR45011-1GEA
16370
CZK
APOSOR45011-1G
APOSOR45011-250MG
APOSOR45011-5G
6-Methylbicyclo[4.1.0]heptan-2-one 95%
6-Methylbicyclo[4.1.0]heptan-2-one
Vzorec:
C₈H₁₂O MW: 124,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09866569 CAS číslo: 14845-41-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...