1,2,4-Triazole-3,4-diamine hydrochloride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR450109-1G
OR450109-250MG
APOSOR450109-1GEA
10350
CZK
APOSOR450109-1G
APOSOR450109-250MG
1,2,4-Triazole-3,4-diamine hydrochloride
1,2,4-Triazole-3,4-diamine hydrochloride
Vzorec:
C₂H₅N₅·ClH MW: 135,556 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11500152 CAS číslo: 6421-05-2 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...