2-Amino-7-hydroxy[1,2,4]triazolo[1,5-a]pyrimidin-5(1H)-one
Dodavatel: Apollo Scientific
OR450107-1G
OR450107-250MG
APOSOR450107-1GEA
12200
CZK
APOSOR450107-1G
APOSOR450107-250MG
2-Amino-7-hydroxy[1,2,4]triazolo[1,5-a]pyrimidin-5(1H)-one
2-Amino-7-hydroxy[1,2,4]triazolo[1,5-a]pyrimidin-5(1H)-one
Vzorec:
C₅H₅N₅O₂ MW: 167,125 g/mol Storage Temperature: Refrigerator |
CAS číslo:
856864-52-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...