Tisk…

5,7-Dichloro-[1,2,4]triazolo[1,5-a]pyrimidine

Dodavatel: Apollo Scientific
Varování

OR450106-1G OR450106-250MG
APOSOR450106-1GEA 11580 CZK
APOSOR450106-1G APOSOR450106-250MG
5,7-Dichloro-[1,2,4]triazolo[1,5-a]pyrimidine
5,7-Dichloro-[1,2,4]triazolo[1,5-a]pyrimidine
Vzorec: C₅H₂Cl₂N₄
MW: 189,002 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10000845
CAS číslo: 78706-26-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR