Tisk…

Thiazolo[3,2-b][1,2,4]triazol-6(5H)-one

Dodavatel: Apollo Scientific

OR450102-1G OR450102-250MG
APOSOR450102-1GEA 10350 CZK
APOSOR450102-1G APOSOR450102-250MG
Thiazolo[3,2-b][1,2,4]triazol-6(5H)-one
Thiazolo[3,2-b][1,2,4]triazol-6(5H)-one
Vzorec: C₄H₃N₃OS
MW: 141,151 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01651966
CAS číslo: 305848-41-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR