Tisk…

4-Chloro-2-oxo-3H-thiazole-5-carbaldehyde

Dodavatel: Apollo Scientific
Varování

OR450101-1G OR450101-250MG
APOSOR450101-1GEA 12200 CZK
APOSOR450101-1G APOSOR450101-250MG
4-Chloro-2-oxo-3H-thiazole-5-carbaldehyde
4-Chloro-2-oxo-3H-thiazole-5-carbaldehyde
Vzorec: C₄H₂ClNO₂S
MW: 163,582 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00594925
CAS číslo: 55359-96-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR