Tisk…

2-Amino-4,5-dihydro-4-thiazoleacetic acid

Dodavatel: Apollo Scientific

OR450099-1G OR450099-250MG
APOSOR450099-1GEA 14490 CZK
APOSOR450099-1G APOSOR450099-250MG
2-Amino-4,5-dihydro-4-thiazoleacetic acid
2-Amino-4,5-dihydro-4-thiazoleacetic acid
Vzorec: C₅H₈N₂O₂S
MW: 160,194 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03772984
CAS číslo: 103541-11-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR