Tisk…

2-Amino-5-bromo-thiazole-4-carboxylic acid

Dodavatel: Apollo Scientific

OR450098-1G OR450098-250MG
APOSOR450098-1GEA 2530 CZK
APOSOR450098-1G APOSOR450098-250MG
2-Amino-5-bromo-thiazole-4-carboxylic acid
2-Amino-5-bromo-thiazole-4-carboxylic acid
Vzorec: C₄H₃BrN₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD08275696
CAS číslo: 858486-46-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR