4-Iminothiazolidin-2-one
Dodavatel: Apollo Scientific
OR450096-1G
OR450096-250MG
APOSOR450096-1GEA
15770
CZK
APOSOR450096-1G
APOSOR450096-250MG
4-Iminothiazolidin-2-one
4-Iminothiazolidin-2-one
Vzorec:
C₃H₄N₂OS Storage Temperature: Ambient |
MDL Number:
MFCD00154755 CAS číslo: 19967-65-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...