Tisk…

Thiazolidine-2,4-dithione

Dodavatel: Apollo Scientific

OR450095-1G OR450095-250MG
APOSOR450095-1GEA 19440 CZK
APOSOR450095-1G APOSOR450095-250MG
Thiazolidine-2,4-dithione
Thiazolidine-2,4-dithione
Vzorec: C₃H₃NS₃
Storage Temperature: Ambient
MDL Number: MFCD06253950
CAS číslo: 4303-27-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR