Tisk…

7-Bromo-6-methoxy-5-azaindole 95+%

Dodavatel: Apollo Scientific

OR450015-100MG OR450015-250MG
APOSOR450015-100MGEA 1540 CZK
APOSOR450015-100MG APOSOR450015-250MG
7-Bromo-6-methoxy-5-azaindole 95+%
7-Bromo-6-methoxy-5-azaindole
Vzorec: C₈H₇BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD12962570
CAS číslo: 1190314-42-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR