[5-(Fur-2-yl)-1H-pyrazol-3-yl]methanol 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-[3-(Hydroxymethyl)-1H-pyrazol-5-yl]furan, 5-(Fur-2-yl)-3-(hydroxymethyl)-1H-pyrazole
OR4420-100MG
OR4420-1G
OR4420-250MG
APOSOR4420-100MGEA
1740
CZK
APOSOR4420-100MG
APOSOR4420-1G
APOSOR4420-250MG
[5-(Fur-2-yl)-1H-pyrazol-3-yl]methanol 97%
[5-(Fur-2-yl)-1H-pyrazol-3-yl]methanol
Vzorec:
C₈H₈N₂O₂ MW: 164,161 g/mol Bod tání: 159 °C Storage Temperature: Ambient |
MDL Number:
MFCD07368534 CAS číslo: 84978-67-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...