Tisk…

1-(2,3-Dihydro-1-benzofuran-5-yl)ethanol

Dodavatel: Apollo Scientific
Varování

OR43687-1G OR43687-250MG
APOSOR43687-1GEA 10150 CZK
APOSOR43687-1G APOSOR43687-250MG
1-(2,3-Dihydro-1-benzofuran-5-yl)ethanol
1-(2,3-Dihydro-1-benzofuran-5-yl)ethanol
Vzorec: C₁₀H₁₂O₂
MW: 164,201 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12047714
CAS číslo: 7228-49-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR