Tisk…

4-[(tert-Butoxycarbonyl)amino]but-2-ynoic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Aminobut-2-ynoic acid, N-BOC protected

OR43679-250MG OR43679-500MG
APOSOR43679-250MGEA 10150 CZK
APOSOR43679-250MG APOSOR43679-500MG
4-[(tert-Butoxycarbonyl)amino]but-2-ynoic acid
4-[(tert-Butoxycarbonyl)amino]but-2-ynoic acid
Vzorec: C₉H₁₃NO₄
MW: 199,204 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11847128
CAS číslo: 168762-94-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR