Tisk…

3-Bromopyrrolidin-2-one

Dodavatel: Apollo Scientific
Varování

OR43677-1G OR43677-100MG OR43677-250MG
APOSOR43677-1GEA 32440 CZK
APOSOR43677-1G APOSOR43677-100MG APOSOR43677-250MG
3-Bromopyrrolidin-2-one
3-Bromopyrrolidin-2-one
Vzorec: C₄H₆BrNO
Storage Temperature: Freezer
MDL Number: MFCD18886531
CAS číslo: 40557-20-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR