4-Methoxybenzenesulphonohydrazide 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-(Hydrazinosulphonyl)anisole, 4-Methoxybenzenesulphonic acid hydrazide
OR43661-25G
OR43661-5G
OR43661-100G
APOSOR43661-25GEA
4140
CZK
APOSOR43661-25G
APOSOR43661-5G
APOSOR43661-100G
4-Methoxybenzenesulphonohydrazide 97%
4-Methoxybenzenesulphonohydrazide
Vzorec:
C₇H₁₀N₂O₃S MW: 202.231 g/mol Bod tání: 109 °C (rozkl.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00014751 CAS číslo: 1950-68-1 EINECS: 217-769-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...