Tisk…

5-Bromo-2-(hydroxymethyl)-1,3-thiazole 95%

Dodavatel: Apollo Scientific

OR43658-250MG OR43658-1G
APOSOR43658-250MGEA 1310 CZK
APOSOR43658-250MG APOSOR43658-1G
5-Bromo-2-(hydroxymethyl)-1,3-thiazole 95%
5-Bromo-2-(hydroxymethyl)-1,3-thiazole
Vzorec: C₄H₄BrNOS
Storage Temperature: Refrigerator
MDL Number: MFCD21728921
CAS číslo: 911052-85-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR