Tisk…

6-Bromo-7-methylquinoline

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 6-Bromo-7-methyl-1-azanaphthalene

OR43652-5G
APOSOR43652-5GEA 5130 CZK
APOSOR43652-5G
6-Bromo-7-methylquinoline
6-Bromo-7-methylquinoline
Vzorec: C₁₀H₈BrN
MW: 222,081 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18260428
CAS číslo: 122759-89-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR