Tisk…

(1H-Indazol-4-yl)acetic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-(1H-Indazol-4-yl)ethanoic acid, 2-(1H-Indazol-4-yl)acetic acid, 4-(Carboxymethyl)-1H-indazole

OR43628-250MG OR43628-1G
APOSOR43628-250MGEA 7910 CZK
APOSOR43628-250MG APOSOR43628-1G
(1H-Indazol-4-yl)acetic acid
(1H-Indazol-4-yl)acetic acid
Vzorec: C₉H₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD13183061
CAS číslo: 902131-33-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR