Tisk…

2-Iodo-6-methoxyaniline

Dodavatel: Apollo Scientific
Varování

Synonyms: 6-Iodo-o-anisidine, 2-Amino-3-iodoanisole

OR43627-1G OR43627-250MG
APOSOR43627-1GEA 1870 CZK
APOSOR43627-1G APOSOR43627-250MG
2-Iodo-6-methoxyaniline
2-Iodo-6-methoxyaniline
Vzorec: C₇H₈INO
MW: 249.049 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11977372
CAS číslo: 354574-31-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR