Tisk…

2-Amino-6-iodophenol 96%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Hydroxy-3-iodoaniline

OR43626-250MG
APOSOR43626-250MGEA 870 CZK
APOSOR43626-250MG
2-Amino-6-iodophenol 96%
2-Amino-6-iodophenol
Vzorec: C₆H₆INO
MW: 235,022 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16999401
CAS číslo: 99968-81-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR