[(4-Methylphenyl)thio]acetic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR4248-5G
OR4248-10G
APOSOR4248-5GEA
910
CZK
APOSOR4248-5G
APOSOR4248-10G
[(4-Methylphenyl)thio]acetic acid 98%
[(4-Methylphenyl)thio]acetic acid
Vzorec:
C₉H₁₀O₂S Bod tání: 79…83 °C Storage Temperature: Ambient |
MDL Number:
MFCD00034675 CAS číslo: 3996-29-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...