Tisk…

Methyl 4-(4-bromophenyl)-4-oxobutanoate 97%

Dodavatel: Apollo Scientific
Varování

OR42289-1G OR42289-250MG OR42289-5G
APOSOR42289-1GEA 3580 CZK
APOSOR42289-1G APOSOR42289-250MG APOSOR42289-5G
Methyl 4-(4-bromophenyl)-4-oxobutanoate 97%
Methyl 4-(4-bromophenyl)-4-oxobutanoate
Vzorec: C₁₁H₁₁BrO₃
MW: 271,107 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03090515
CAS číslo: 30913-86-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR