2-Amino-4-cyclopropylpyridine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Cyclopropylpyridin-2-amine
OR42284-1G
OR42284-250MG
OR42284-5G
APOSOR42284-1GEA
5560
CZK
APOSOR42284-1G
APOSOR42284-250MG
APOSOR42284-5G
2-Amino-4-cyclopropylpyridine
2-Amino-4-cyclopropylpyridine
Vzorec:
C₈H₁₀N₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD12032793 CAS číslo: 908269-97-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...