Tisk…

1-(5-Bromo-1-benzofuran-2-yl)ethanol

Dodavatel: Apollo Scientific
Varování

OR42283-1G OR42283-250MG OR42283-5G
APOSOR42283-1GEA 3740 CZK
APOSOR42283-1G APOSOR42283-250MG APOSOR42283-5G
1-(5-Bromo-1-benzofuran-2-yl)ethanol
1-(5-Bromo-1-benzofuran-2-yl)ethanol
Vzorec: C₁₀H₉BrO₂
MW: 241,081 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08443121
CAS číslo: 592542-03-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR