5-Bromo-4-methyl-2-(methylsulphonyl)pyridine
Dodavatel: Apollo Scientific
|
Varování
|
OR42264-1G
OR42264-500MG
APOSOR42264-1GEA
6780
CZK
APOSOR42264-1G
APOSOR42264-500MG
5-Bromo-4-methyl-2-(methylsulphonyl)pyridine
5-Bromo-4-methyl-2-(methylsulphonyl)pyridine
Vzorec:
C₇H₈BrNO₂S Storage Temperature: Ambient |
MDL Number:
MFCD28397882 CAS číslo: 1279106-02-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...