6-Chloroquinoline-3-carboxylic acid
Dodavatel: Apollo Scientific
OR42243-1G
OR42243-5G
OR42243-250MG
APOSOR42243-1GEA
2700
CZK
APOSOR42243-1G
APOSOR42243-5G
APOSOR42243-250MG
6-Chloroquinoline-3-carboxylic acid
6-Chloroquinoline-3-carboxylic acid
Vzorec:
C₁₀H₆ClNO₂ Bod tání: 228 °C Storage Temperature: Ambient |
MDL Number:
MFCD09787826 CAS číslo: 118791-14-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...