1-Bromo-3-(3-bromoprop-1-ynyl)benzene 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
1-Bromo-3-(3-bromoprop-1-yn-1-yl)benzene
OR42053-1G
OR42053-250MG
OR42053-5G
APOSOR42053-1GEA
2650
CZK
APOSOR42053-1G
APOSOR42053-250MG
APOSOR42053-5G
1-Bromo-3-(3-bromoprop-1-ynyl)benzene 98%
1-Bromo-3-(3-bromoprop-1-ynyl)benzene
Vzorec:
C₉H₆Br₂ MW: 273,952 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD22482458 CAS číslo: 1333171-42-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...