1-[(1-Methylpiperidin-3-yl)methyl]piperazine 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR4163-10G
OR4163-2G
APOSOR4163-10GEA
10520
CZK
APOSOR4163-10G
APOSOR4163-2G
1-[(1-Methylpiperidin-3-yl)methyl]piperazine 97%
1-[(1-Methylpiperidin-3-yl)methyl]piperazine
Vzorec:
C₁₁H₂₃N₃ MW: 197,32 g/mol Bod varu: 75…76 °C (0,25 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD02093538 CAS číslo: 496808-03-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...