4-[(But-1-ylcarbamoyl)amino]benzeneboronic acid, pinacol ester
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-(3-But-1-ylureido)benzeneboronic acid, pinacol ester
OR4122-1G
OR4122-5G
OR4122-10G
APOSOR4122-1GEA
4370
CZK
APOSOR4122-1G
APOSOR4122-5G
APOSOR4122-10G
4-[(But-1-ylcarbamoyl)amino]benzeneboronic acid, pinacol ester
4-[(But-1-ylcarbamoyl)amino]benzeneboronic acid, pinacol ester
Vzorec:
C₁₇H₂₇BN₂O₃ MW: 318,219 g/mol Bod tání: 207…209 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD07363843 CAS číslo: 850567-59-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...