3-Bromo-2,2-bis(bromomethyl)propanoic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR41167-5G
OR41167-100G
APOSOR41167-5GEA
5590
CZK
APOSOR41167-5G
APOSOR41167-100G
3-Bromo-2,2-bis(bromomethyl)propanoic acid
3-Bromo-2,2-bis(bromomethyl)propanoic acid
Vzorec:
C₅H₇Br₃O₂ Bod tání: 97…99 °C Storage Temperature: Ambient |
MDL Number:
MFCD00128769 CAS číslo: 52813-48-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...