3-(Chloromethyl)-4-methyl-1H-1,2,4-triazol-5(4H)-one 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR41033-1G
OR41033-250MG
APOSOR41033-1GEA
6550
CZK
APOSOR41033-1G
APOSOR41033-250MG
3-(Chloromethyl)-4-methyl-1H-1,2,4-triazol-5(4H)-one 98%
3-(Chloromethyl)-4-methyl-1H-1,2,4-triazol-5(4H)-one
Vzorec:
C₄H₆ClN₃O MW: 147,563 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD22741615 CAS číslo: 1338226-21-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...